Tisk…
Hydrogen generator, Precision Hydrogen 1200
Dodavatel: PEAK SCIENTIFIC
3300843
PEAK3300843EA 504040 CZK
PEAK3300843
Hydrogen generator, Precision Hydrogen 1200
Generátory plynu Generátory vodíku
The Precision Hydrogen 1200 is the first 1L+ Hydrogen generator and it is designed to produce hydrogen at a flow of 1200 ml with up to 99,9995% purity. Precision 1200 H₂ generator provides the gas needed for detectors requiring Hydrogen fuel gas, such as FID, NPD and SCD, and it can also support GC carrier gases. One generator is capable of supplying multiple detectors, and is available in various flow rates to ideally suit individual customers’ needs.
  • Internal leak detection with automatic shutdown features
  • Automatic loading pump as standard
  • Simple
More Product Information