Tisk…
Shelving units
Dodavatel: Palbam
SU-1340-4-P SU-1240-4P SU-1060-4-P SU-1060-4-S SU-1040-4-S SU-1040-4-S-C SU-1060-4-S-C SU-1640-4-P SU-1640-4-S SU1660-S-SHELF
132-1060EA 0 CZK
132-1060 132-1059 132-1056 132-1057 132-0998 132-0999 132-1058 PALBSU-1640-4-P PALBSU-1640-4-S PALBSU1660-S-SHELF
Shelving units
Nábytek kancelářský Police
High-quality cleanroom shelving units on leveller legs with height adjustable feet or on white nylon 130 mm (5") cleanroom casters.
  • Shelf options: Solid or perforated
  • Shelves are removable or height adjustable every 200 mm (8")
  • 4 shelves per unit (extra shelves can be ordered separately)
Stainless steel 304, electropolished.