Tisk…
SEAP assay kit
Dodavatel: OZ BIOSCIENCES
SP00500
OZBISP00500EA 4360 CZK
OZBISP00500
SEAP assay kit
Test Buněčná stanovení
SEAP assay kit (Secreted Alkaline Phosphatase) is a colorimetric assay for sensitive quantification of SEAP in culture medium from transfected cells or tissues.
  • Designed to detect SEAP reporter gene expression
  • Fast and easy analysis
  • Colorimetric-measure with a standard spectrophotometer or an ELISA reader (405 nm)
  • Multiple samples can be analysed in low volumes
  • Convenient and economical packaging: no tablet, no powder
Alkaline phosphatase cleaves the colourless 4-nitrophenylphosphate (pNPP) substrate to give a yellow p-nitrophenolate product in alkaline solutions. It is commonly used as a reporter gene expression and can be accurately quantified. It is a truncated form of placental alkaline phosphatase (GPI-anchored protein).
Informace o dodávce: Kit contains pNPP-containing assay buffer component A (10×1 ml), reaction buffer component B (125 ml), reaction buffer component C (125 µl) and dilution buffer component D (50 ml).
Upozornení: Store at +4 °C.