Tisk…
Ag-AK, Immunofluorescence test, IMAGEN™
Dodavatel: THERMO FISHER DIAGNOSTICS
S610830-2 S610930-2 S611030-2 S611130-2 S611230-2 S611330-2 S611430-6
OXOIS610830-2EA 4620 CZK
OXOIS610830-2 OXOIS610930-2 OXOIS611030-2 OXOIS611130-2 OXOIS611230-2 OXOIS611330-2 OXOIS611430-6
Ag-AK, Immunofluorescence test, IMAGEN™
Mikrobiologické testovací systémy Antigenové Protilátkové Testy AK-AK, imunotest
IMAGEN™ is a qualitative direct immunofluorescence test used for detection and confirmation of the presence of virus in cell culture or in clinical specimen.
  • Very specific