Tisk…
Antimicrobial susceptibility disks
Dodavatel: THERMO FISHER DIAGNOSTICS
CT0002B CT0003B CT0005B CT0007B CT0010B CT0011B CT0013B CT0015B CT0016B CT0017B CT0018B CT0019B CT0020B CT0021B CT0023B CT0024B CT0025B CT0026B CT0028B CT0030B CT0031B CT0032B CT0033B CT0034B CT0036B CT0037B CT0038B CT0040B CT0041B CT0042B CT0043B CT0044B CT0046B CT0047B CT0048B CT0051B CT0052B CT0054B CT0056B CT0057B CT0058B CT0059B CT0060B CT0061B CT0063B CT0064B CT0067B CT0069B CT0072B CT0076B CT0088B CT0091B CT0096B CT0104B CT0107B CT0119B CT0123B CT0124B CT0127B CT0149B CT0152B CT0159B CT0161B CT0166B CT0167B CT0180B CT0183B CT0188B CT0193B CT0199B CT0207B CT0223B CT0225B CT0232B CT0249B CT0263B CT0264B CT0407B CT0411B CT0412B CT0417B CT0424B CT0425B CT0434B CT0446B CT0453B CT0455B CT0466B CT0493B CT0520B CT0523B CT0538B CT0545B CT0623B CT0639B CT0647B CT0653B CT0661B CT0664B CT0665B CT0666B CT0668B CT0693B CT0725B CT0758B CT0774B CT0794B CT0998B CT0771B CT0906B CT1587B CT1612B CT1629B CT1633B CT1650B CT1662B CT1751B CT1754B CT1756B CT1761B CT1806B CT1807B CT0823B CT1616B CT1841B CT1880B CT1897B CT1758B
OXOICT0002BEA 1540 CZK
OXOICT0002B OXOICT0003B OXOICT0005B OXOICT0007B OXOICT0010B OXOICT0011B OXOICT0013B OXOICT0015B OXOICT0016B OXOICT0017B OXOICT0018B OXOICT0019B OXOICT0020B OXOICT0021B OXOICT0023B OXOICT0024B OXOICT0025B OXOICT0026B OXOICT0028B OXOICT0030B OXOICT0031B OXOICT0032B OXOICT0033B OXOICT0034B OXOICT0036B OXOICT0037B OXOICT0038B OXOICT0040B OXOICT0041B OXOICT0042B OXOICT0043B OXOICT0044B OXOICT0046B OXOICT0047B OXOICT0048B OXOICT0051B OXOICT0052B OXOICT0054B OXOICT0056B OXOICT0057B OXOICT0058B OXOICT0059B OXOICT0060B OXOICT0061B OXOICT0063B OXOICT0064B OXOICT0067B OXOICT0069B OXOICT0072B OXOICT0076B OXOICT0088B OXOICT0091B OXOICT0096B OXOICT0104B OXOICT0107B OXOICT0119B OXOICT0123B OXOICT0124B OXOICT0127B OXOICT0149B OXOICT0152B OXOICT0159B OXOICT0161B OXOICT0166B OXOICT0167B OXOICT0180B OXOICT0183B OXOICT0188B OXOICT0193B OXOICT0199B OXOICT0207B OXOICT0223B OXOICT0225B OXOICT0232B OXOICT0249B OXOICT0263B OXOICT0264B OXOICT0407B OXOICT0411B OXOICT0412B OXOICT0417B OXOICT0424B OXOICT0425B OXOICT0434B OXOICT0446B OXOICT0453B OXOICT0455B OXOICT0466B OXOICT0493B OXOICT0520B OXOICT0523B OXOICT0538B OXOICT0545B OXOICT0623B OXOICT0639B OXOICT0647B OXOICT0653B OXOICT0661B OXOICT0664B OXOICT0665B OXOICT0666B OXOICT0668B OXOICT0693B OXOICT0725B OXOICT0758B OXOICT0774B OXOICT0794B OXOICT0998B OXOICT0771B OXOICT0906B OXOICT1587B OXOICT1612B OXOICT1629B OXOICT1633B OXOICT1650B OXOICT1662B OXOICT1751B OXOICT1754B OXOICT1756B OXOICT1761B OXOICT1806B OXOICT1807B OXOICT0823B OXOICT1616B OXOICT1841B OXOICT1880B OXOICT1897B OXOICT1758B
Antimicrobial susceptibility disks
Mikrobiologické testovací systémy Antibiotické Testy
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use antimicrobial susceptibility disks (5×50).
  • Reagent code and concentration printed clearly on each disk
  • Shatterproof, spring-loaded cartridges for easy dispensing
  • Cartridges individually blister-packed with desiccant to maintain product quality