Tisk…
MinION™ Mk1D DNA/RNA Sequencing Device
Dodavatel: Oxford Nanopore Technologies
SUPP1D MDP-RPB114 BASIC12M-MK1D MDIP-LSK114 MDIP-NBD14.24 MDIP-PCS14 MDIP-RAD14 MDIP-RBK14.24 MDIP-RNA MDIP-RPB114 MDP-LSK114 MDP-NBD14.24 MDP-PCS14 MDP-RAD14 MDP-RBK14.24 MDP-RNA
OXNTSUPP1DEA 164880 CZK
OXNTSUPP1D OXNTMDP-RPB114 OXNTBASIC12M-MK1D OXNTMDIP-LSK114 OXNTMDIP-NBD14.24 OXNTMDIP-PCS14 OXNTMDIP-RAD14 OXNTMDIP-RBK14.24 OXNTMDIP-RNA OXNTMDIP-RPB114 OXNTMDP-LSK114 OXNTMDP-NBD14.24 OXNTMDP-PCS14 OXNTMDP-RAD14 OXNTMDP-RBK14.24 OXNTMDP-RNA
MinION™ Mk1D DNA/RNA Sequencing Device
Next Generation Sequencing Instruments
MinION™ - your personal, portable DNA and RNA sequencer.
  • Any length read from short to ultra long: 20 bp to >4 Mb
  • High-accuracy, direct sequencing of native DNA and RNA with built-in modification and best-in-class methylation detection
  • Real-time sequencing for immediate access to actionable results
  • Simple 10-minute sample prep available
  • Portable, pocket-sized device sequence in the field or lab
A sequencer for everyone - empowering individual researchers, labs and those new to DNA/RNA sequencing to perform in-house sequencing and take control ofMore Product Information
New Product
Objednat
Produktová specifikace
Environmentally Preferable
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance.