Tisk…
Short Fragment Eliminator Expansion
Dodavatel: Oxford Nanopore Technologies
EXP-SFE001
OXNTEXP-SFE001EA 2770 CZK
OXNTEXP-SFE001
Short Fragment Eliminator Expansion
Činidla pro nukleové kyseliny Next Generation Sequencing Reagents
An expansion pack designed for size selection of high molecular weight DNA samples to progressively deplete short fragments of <25 kb.
  • Size select to progressively deplete short fragments (<25 kb) prior to sequencing library preparation
  • Enrich sequencing libraries for long fragments without using any external third-party reagents
The short fragment eliminator (EXP-SFE001) expansion pack is recommended for users who want to size select high molecular-weight (HMW) gDNA to deplete short fragments (<25 kb) using size selective precipitation. The kit contains Short Fragment Eliminator (SFE) buffer which progressively removes fragments up to 25 kb, with fragments under 10 kb being almost completely removed in a quick and simple workflow.
NOTE: This kit is designed to remove short DNA fragments and requires samples to be predominantly made up of HMW DNA (>48 kb). If there are large amounts of short fragments in your sample, this will result in a low yield at the end of the prep and a low sequencing output.
Informace o dodávce: Kits are shipped at 2 to 8 °C in an environmentally-friendly temperature-controlled shipping box. Upon receipt, please place the product in a long-term storage location at −20 °C. Oxford Nanopore Technologies deem the useful life of the product to be 3 months from receipt by the customer.
Objednat
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance.