Tisk…
ORP standards for Redox/ORP electrodes, Orion™
Dodavatel: Thermo Orion
967961 967901
ORIO967961EA 3310 CZK
ORIO967961 ORIO967901
ORP standards for Redox/ORP electrodes, Orion™
Standardy Standardy redox/ORP
Measure ORP simply and accurately with Thermo Scientific™ Orion™ ORP standards for Redox/ORP electrodes
  • Does not contain hazardous chemicals
  • Included in the new ASTM ORP method
  • Used as relative mV standard to calibrate the offset of sample
Orion™ OPR standards are designed for use with ORP/ATC Triode™ electrodes and Sure-Flow™ ORP electrodes.
Stable for up to a year if unopened and can be used directly from the bottle with no mixing or diluting.