Tisk…
pH/mV/°C/ISE meter, bench, Orion™ Dual Star™
Dodavatel: Thermo Orion
2115000 2115001 2115101 2115102 2115205 2115203 2115202 2115204 2115201
ORIO2115000EA 64040 CZK
ORIO2115000 ORIO2115001 ORIO2115101 ORIO2115102 ORIO2115205 ORIO2115203 ORIO2115202 ORIO2115204 ORIO2115201
pH/mV/°C/ISE meter, bench, Orion™ Dual Star™
Meřiče elektrochemické Meřiče pH a multimetry
These bench top meters with two measuring channels are designed for accurate and reliable pH, ion concentration, mV, ORP and temperature measurements. Ideal for high throughput pH and ion analysis in the lab.
  • Numeric keypad and menu-specific function keys; split screen display for viewing two measurements at the same time
  • Perform up to a 6 point pH calibration with automatic recognition for USA/NIST and EURO/DIN buffers with option to manually input custom buffer values
  • Perform up to a 6 point ion concentration (ISE) calibration with non-linear auto-blank
More Product Information