Tisk…

Anti-KRAS Mouse Monoclonal Antibody [clone: OTI2F8]

Dodavatel: OriGene
TA801227
ORIGTA801227EA 17690 CZK
ORIGTA801227
Anti-KRAS Mouse Monoclonal Antibody [clone: OTI2F8]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: KRAS (Kirsten rat sarcoma viral oncogene homolog)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR