Tisk…

Anti-RIPK1 Mouse Monoclonal Antibody [clone: OTI2G3]

Dodavatel: OriGene
TA800325
ORIGTA800325EA 17690 CZK
ORIGTA800325
Anti-RIPK1 Mouse Monoclonal Antibody [clone: OTI2G3]
Protilátky
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150

Type: Primary
Antigen: RIPK1 (receptor (TNFRSF)-interacting serine-threonine kinase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR