Tisk…

Anti-CTBS Rabbit Polyclonal Antibody

Dodavatel: OriGene
TA590635
ORIGTA590635EA 19050 CZK
ORIGTA590635
Anti-CTBS Rabbit Polyclonal Antibody
Protilátky
Recommended Dilutions: Western Blot 1:20000, IHC 1:150, ELISA 1:100-1:2000

Type: Primary
Antigen: CTBS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Produktová specifikace

Symbol antigenu CTBS
Environmentally Preferable
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Human
Hostitel Rabbit
Izotyp IgG
Western blot Yes
ELISA Yes
Imunochemie Yes
Imunogen DNA immunization. This antibody was made against a protein fragment from the C Terminus Region
Formulář Concentrated
Čištění The fragment used for DNA immunization was expressed in E.coli and the purified protein fragment was used for affinity purification of the antibody.
Skladovací pufr 20 mM Potassium Phosphate, 150 mM Sodium Chloride, pH 7.0
Skladovací teplota –20 °C
Transportní teplota –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR