Tisk…

Anti-RASSF8 Mouse Monoclonal Antibody [clone: OTI1A1]

Dodavatel: OriGene
TA505927
ORIGTA505927EA 17690 CZK
ORIGTA505927
Anti-RASSF8 Mouse Monoclonal Antibody [clone: OTI1A1]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150-510

Type: Primary
Antigen: RASSF8 (Ras association (RalGDS/AF-6) domain family (N-terminal) member 8)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR