Tisk…

Anti-BTLA Mouse Monoclonal Antibody [clone: OTI2E4]

Dodavatel: OriGene
TA505688
ORIGTA505688EA 17690 CZK
ORIGTA505688
Anti-BTLA Mouse Monoclonal Antibody [clone: OTI2E4]
Protilátky
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: BTLA (B and T lymphocyte associated)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR