Tisk…

Anti-TDO2 Mouse Monoclonal Antibody [clone: OTI4G2]

Dodavatel: OriGene
TA504730
ORIGTA504730EA 17690 CZK
ORIGTA504730
Anti-TDO2 Mouse Monoclonal Antibody [clone: OTI4G2]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: TDO2 (tryptophan 2,3-dioxygenase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR