Tisk…

Anti-RASD2 Mouse Monoclonal Antibody [clone: OTI1A12]

Dodavatel: OriGene
TA502070
ORIGTA502070EA 17690 CZK
ORIGTA502070
Anti-RASD2 Mouse Monoclonal Antibody [clone: OTI1A12]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: RASD2 (RASD family, member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR