Tisk…

Anti-BIRC5 Mouse Monoclonal Antibody [clone: OTI2D9]

Dodavatel: OriGene
TA501245
ORIGTA501245EA 17690 CZK
ORIGTA501245
Anti-BIRC5 Mouse Monoclonal Antibody [clone: OTI2D9]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: BIRC5 (baculoviral IAP repeat containing 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR