Tisk…

Anti-CD36 Mouse Monoclonal Antibody [clone: OTI3F4]

Dodavatel: OriGene
TA500921
ORIGTA500921EA 17690 CZK
ORIGTA500921
Anti-CD36 Mouse Monoclonal Antibody [clone: OTI3F4]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: CD36 (CD36 molecule (thrombospondin receptor))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR