Tisk…

Anti-ANGPTL3 Rabbit Polyclonal Antibody

Dodavatel: OriGene
TA321837
ORIGTA321837EA 15040 CZK
ORIGTA321837
Anti-ANGPTL3 Rabbit Polyclonal Antibody
Protilátky
Recommended Dilutions: ELISA: 1:1000-5000; Western Blot: 1:500-2000; Immunochemistry: 1:25-100

Type: Primary
Antigen: ANGPTL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR