Tisk…

Anti-Pck1 Rabbit Polyclonal Antibody

Dodavatel: OriGene
TA319014
ORIGTA319014EA 19140 CZK
ORIGTA319014
Anti-Pck1 Rabbit Polyclonal Antibody
Protilátky
Recommended Dilutions: Western Blot: 0.5-4 ?g/ml

Type: Primary
Antigen: Pck1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR