Tisk…

Anti-Cfd Rabbit Polyclonal Antibody

Dodavatel: OriGene
TA319009
ORIGTA319009EA 16090 CZK
ORIGTA319009
Anti-Cfd Rabbit Polyclonal Antibody
Protilátky
Recommended Dilutions: Western Blot: 0.5-4 ?g/ml

Type: Primary
Antigen: Cfd
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR