Tisk…

Anti-KDM3A Rabbit Polyclonal Antibody

Dodavatel: OriGene
TA306828
ORIGTA306828EA 22550 CZK
ORIGTA306828
Anti-KDM3A Rabbit Polyclonal Antibody
Protilátky
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: KDM3A (lysine (K)-specific demethylase 3A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR