Tisk…

Anti-VPS35 Goat Polyclonal Antibody

Dodavatel: OriGene
TA302699
ORIGTA302699EA 22360 CZK
ORIGTA302699
Anti-VPS35 Goat Polyclonal Antibody
Protilátky
Recommended Dilutions: ELISA: 1:32000; Western Blot: 0.01-0.1?g/ml

Type: Primary
Antigen: VPS35 (vacuolar protein sorting 35 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR