Tisk…
Mouse Untagged Clone Plasmid, Lrrc15 (NM_028973)
Dodavatel: OriGene
MC219256
ORIGMC219256EA 32180 CZK
ORIGMC219256
Mouse Untagged Clone Plasmid, Lrrc15 (NM_028973)
Vektory, plazmidy a knihovny
Lrrc15 (untagged) - Mouse leucine rich repeat containing 15 (Lrrc15).
  • Centrifuge at 5,000×g for 5 mins
  • Carefully open the tube and add 100ul of sterile water to dissolve the DNA
  • Close the tube and incubate for 10 minutes at room temperature
  • Briefly vortex the tube and then do a quick spin (less than 5000×g) to concentrate the liquid at the bottom
  • Store the suspended plasmid at −20 °C
  • The DNA is stable for at least one year from date of shipping when stored at −20 °C
The ORF clone is ion-exchange column purified and shipped in a 2D barcoded Matrix tube containing 10ug of transfection-ready, dried plasmid DNA (reconstitute with 100 µl of water).
The molecular sequence of this clone aligns with the gene accession number as a point of reference only. However, individual transcript sequences of the same gene can differ through naturally occurring variations (e.g. polymorphisms), each with its own valid existence.