Tisk…

Anti-PEG3 Mouse Monoclonal Antibody [clone: 1B2]

Dodavatel: OriGene
CF801958
ORIGCF801958EA 23720 CZK
ORIGCF801958
Anti-PEG3 Mouse Monoclonal Antibody [clone: 1B2]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PEG3
Clonality: Monoclonal
Clone: 1B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR