Tisk…

Anti-PLEK Mouse Monoclonal Antibody [clone: 2D11]

Dodavatel: OriGene
CF500900
ORIGCF500900EA 23720 CZK
ORIGCF500900
Anti-PLEK Mouse Monoclonal Antibody [clone: 2D11]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Flow Cytometry: 1:100

Type: Primary
Antigen: PLEK
Clonality: Monoclonal
Clone: 2D11
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR