Tisk…
Omnifit® EZ SolventPlus™ Glass Chromatography Columns for FPLC and LPLC
Dodavatel: OMNIFIT
006EZS-10-25AA 006EZS-10-25AF 006EZS-50-25AF 006EZS-50-25AA
OMNI006EZS-10-25AAEA 0 CZK
OMNI006EZS-10-25AA OMNI006EZS-10-25AF OMNI006EZS-50-25AF OMNI006EZS-50-25AA
Omnifit® EZ SolventPlus™ Glass Chromatography Columns for FPLC and LPLC
Kolony chromatografické
EZ SolventPlus™ columns are designed for maximum chemical compatibility. Borosilicate glass and PTFE construction, with Chemraz® o-rings and PTFE frits, only the most inert materials are present in the fluid path. ETFE end caps and fitting nuts ensure a chemically inert connection into and out of the system.
  • Available in six diameters and a variety of lengths
  • Operating temperature: 4 to 40 °C
  • Glass column is made up of borosilicate glass
Designed for techniques using organic solvents such as reversed phase chromatography, but equally compatible withMore Product Information