Tisk…
COURIER™ 5000 Shipping Scales
Dodavatel: OHAUS
30745890 30745891 30745892 30745893 30745894 30745895 30745896 30820402 30820403 30820404 30820405 30820408 30820406 30820407
OHAU30745890EA 9990 CZK
OHAU30745890 OHAU30745891 OHAU30745892 OHAU30745893 OHAU30745894 OHAU30745895 OHAU30745896 OHAU30820402 OHAU30820403 OHAU30820404 OHAU30820405 OHAU30820408 OHAU30820406 OHAU30820407
COURIER™ 5000 Shipping Scales
Váhy
The Courier 5000 series shipping scales feature a sturdy low-profile design with stainless steel top, making them tough enough for shipping applications and easy to clean for small veterinary applications.
  • For weighing, dynamic weighing/display hold, weight alert, totalisation
  • 5-digit, 7-segment LCD display with white backlight, 28 mm-high digits
  • Standard AC adapter or USB type C cable included
  • Standard USB type C communication
  • ABS indicator, 304 stainless steel platform, powder-coated frame
  • 150% overload protection
Data is clearly legible on a large,More Product Information