Tisk…
Heating blocks for Ohaus block heaters
Dodavatel: OHAUS
30400190 30400189 30400160 30400161 30400164 30400166 30400169 30400170 30400171 30400182 30400183 30400184 30400185 30400186 30400187 30400188
OHAU30400190EA 4230 CZK
OHAU30400190 OHAU30400189 OHAU30400160 OHAU30400161 OHAU30400164 OHAU30400166 OHAU30400169 OHAU30400170 OHAU30400171 OHAU30400182 OHAU30400183 OHAU30400184 OHAU30400185 OHAU30400186 OHAU30400187 OHAU30400188
Heating blocks for Ohaus block heaters
Termostaty blokové Bloky
Solid blocks: For use as a low temperature hotplate or for custom drilling.
Blocks for cuvettes: Two parallel slots accommodate 6 cuvettes in each slot, side by side for excellent stability and heat transfer.
Blocks for PCR -plates, tubes, strips: Tapered tube wells for 0,2 ml tubes. Spaced for easy access and removal.