Tisk…
Low temperature covers
Dodavatel: OHAUS
30400179 30400180 30400181
OHAU30400179EA 3710 CZK
OHAU30400179 OHAU30400180 OHAU30400181
Low temperature covers
Termostaty blokové Bloky
Plexiglass cover reduces air flow for additional temperature stability in low temperature applications.
  • Two sides are open 6,4 mm