Tisk…
Modular heating blocks
Dodavatel: OHAUS
30400191 30400192 30400193 30400194 30400195 30400151 30400152 30400153 30400154 30400155 30400156 30400157 30400158 30400159 30400162 30400163 30400165 30400167 30400168 30400172
OHAU30400191EA 4230 CZK
OHAU30400191 OHAU30400192 OHAU30400193 OHAU30400194 OHAU30400195 OHAU30400151 OHAU30400152 OHAU30400153 OHAU30400154 OHAU30400155 OHAU30400156 OHAU30400157 OHAU30400158 OHAU30400159 OHAU30400162 OHAU30400163 OHAU30400165 OHAU30400167 OHAU30400168 OHAU30400172
Modular heating blocks
Termostaty blokové Bloky
Modular blocks are constructed from a solid anodized aluminium block. The close contact of tubes-to-block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature.
Upozornení: To avoid possible electrical hazard, do not fill well or block with water or other fluids. Units are designed as a dry bath/incubator.