Tisk…
Industrial scales, modular, Ranger 7000
Dodavatel: OHAUS
30088847 30088844 30088845 30088846 30088834 30088835 30104921 30104922 30104923 30104924 30088848 30088849 30088850 30088851 30104925 30104926 30104927 30104928 30088832 30088833 30104929 30104940 30104941 30104942 30088836 30088837 30088838 30088839 30104943 30104944 30104945 30104946
OHAU30088847EA 43820 CZK
OHAU30088847 OHAU30088844 OHAU30088845 OHAU30088846 OHAU30088834 OHAU30088835 OHAU30104921 OHAU30104922 OHAU30104923 OHAU30104924 OHAU30088848 OHAU30088849 OHAU30088850 OHAU30088851 OHAU30104925 OHAU30104926 OHAU30104927 OHAU30104928 OHAU30088832 OHAU30088833 OHAU30104929 OHAU30104940 OHAU30104941 OHAU30104942 OHAU30088836 OHAU30088837 OHAU30088838 OHAU30088839 OHAU30104943 OHAU30104944 OHAU30104945 OHAU30104946
Industrial scales, modular, Ranger 7000
Váhy
The Ranger 7000 series are rugged, high performance industrial scales for the toughest jobs. Ten advanced application modes can provide results for complex applications while minimising manual calculations. Ranger 7000 will meet a variety of industrial applications from recipes formulation through automatic filling systems to density weighing of concrete. The scales have a bright backlit display and navigation panel that is comprised of a keypad with 5 quick, 8 function, and 12 alphanumeric keys and a 109 mm graphic display, which can be operated in multiple languages.