Tisk…
Serological pipettes, Nunc™
Dodavatel: Thermo Fisher Scientific
170353N 170354N 170355N 170356N 170357N 170358N 170364N 170365N 170366N 170367N 170368N 170369N 170371N 170372N 170373N 170374N 170375N 170376N
NUNC170353NEA 0 CZK
NUNC170353N NUNC170354N NUNC170355N NUNC170356N NUNC170357N NUNC170358N NUNC170364N NUNC170365N NUNC170366N NUNC170367N NUNC170368N NUNC170369N NUNC170371N NUNC170372N NUNC170373N NUNC170374N NUNC170375N NUNC170376N
Serological pipettes, Nunc™
Pipety Pipety serologické
Stringent quality specifications, and convenient features make Thermo Scientific™ Nunc™ Serological pipettes a great choice for daily lab work. Bold, clear bi-directional graduations make it easy-to-read fluid volumes and dispensing amounts.
  • Colour-coded pipettes and packaging for convenient volume selection
  • Reduce packaging waste with bulk packed options
  • PET filter plug to help prevent contamination
  • Clear appearance for better visibility
  • Free of RNases, DNases, and human DNA, non-cytotoxic, non-pyrogenic
  • Ensures dispensing accuracy ±2% for 1 ml and 2 ml
More Product Information
Objednat
Produktová specifikace
Environmentally Preferable Low Manufacturing Impact
Certifikát ochrany životního prostředí NAR Certificate of Retirement