Tisk…

Anti-HMGB1 Mouse Monoclonal Antibody [clone: 19N15F4]

Dodavatel: Novus Biologicals
NBP2-27401
NOVUNBP2-27401EA 12360 CZK
NOVUNBP2-27401
Anti-HMGB1 Mouse Monoclonal Antibody [clone: 19N15F4]
Protilátky
Type: Primary
Antigen: HMGB1 (high mobility group box 1)
Clonality: monoclonal
Clone: 19N15F4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR