Tisk…

Anti-Importin-8 Rabbit Polyclonal Antibody

Dodavatel: Novus Biologicals
NBP2-24751
NOVUNBP2-24751EA 16970 CZK
NOVUNBP2-24751
Anti-Importin-8 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Importin-8
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR