Tisk…

Anti-KDM5B Rabbit Polyclonal Antibody (DyLight® 650)

Dodavatel: Novus Biologicals
NBP1-97310C
NOVUNBP1-97310CEA 20560 CZK
NOVUNBP1-97310C
Anti-KDM5B Rabbit Polyclonal Antibody (DyLight® 650)
Protilátky
Type: Primary
Antigen: KDM5B (lysine (K)-specific demethylase 5B)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR