Tisk…

Anti-ZNF521 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Novus Biologicals
NBP1-91271H
NOVUNBP1-91271HEA 20560 CZK
NOVUNBP1-91271H
Anti-ZNF521 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Type: Primary
Antigen: ZNF521 (zinc finger protein 521)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR