Tisk…

Anti-CHORDC1 Rabbit Polyclonal Antibody (Biotin)

Dodavatel: Novus Biologicals
NBP1-78304B
NOVUNBP1-78304BEA 19020 CZK
NOVUNBP1-78304B
Anti-CHORDC1 Rabbit Polyclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: CHORDC1 (cysteine and histidine-rich domain (CHORD) containing 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR