Tisk…

Anti-HIF1A Rabbit Polyclonal Antibody (DyLight® 405) [clone: exon 10]

Dodavatel: Novus Biologicals
NBP1-47179V
NOVUNBP1-47179VEA 14920 CZK
NOVUNBP1-47179V
Anti-HIF1A Rabbit Polyclonal Antibody (DyLight® 405) [clone: exon 10]
Protilátky
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: polyclonal
Clone: exon 10
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR