Tisk…

Anti-BIRC5 Mouse Monoclonal Antibody (DyLight® 650) [clone: 8E2]

Dodavatel: Novus Biologicals
NB500-644C
NOVUNB500-644CEA 18650 CZK
NOVUNB500-644C
Anti-BIRC5 Mouse Monoclonal Antibody (DyLight® 650) [clone: 8E2]
Protilátky
Type: Primary
Antigen: BIRC5 (baculoviral IAP repeat containing 5)
Clonality: monoclonal
Clone: 800
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR