Tisk…

Anti-HIST3H3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Novus Biologicals
NB21-1081H
NOVUNB21-1081HEA 24920 CZK
NOVUNB21-1081H
Anti-HIST3H3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR