Tisk…

Anti-APOE Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: WUE-4]

Dodavatel: Novus Biologicals
NB110-60531F
NOVUNB110-60531FEA 22950 CZK
NOVUNB110-60531F
Anti-APOE Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: WUE-4]
Protilátky
Type: Primary
Antigen: APOE (apolipoprotein E)
Clonality: monoclonal
Clone: WUE-4
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR