Tisk…

Anti-BRCA1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: MU]

Dodavatel: Novus Biologicals
NB100-600F
NOVUNB100-600FEA 20560 CZK
NOVUNB100-600F
Anti-BRCA1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: MU]
Protilátky
Type: Primary
Antigen: BRCA1 (breast cancer 1, early onset)
Clonality: monoclonal
Clone: MU
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR