Tisk…

Anti-BRCA1 Mouse Monoclonal Antibody (Biotin) [clone: MU]

Dodavatel: Novus Biologicals
NB100-600B
NOVUNB100-600BEA 20560 CZK
NOVUNB100-600B
Anti-BRCA1 Mouse Monoclonal Antibody (Biotin) [clone: MU]
Protilátky
Type: Primary
Antigen: BRCA1 (breast cancer 1, early onset)
Clonality: monoclonal
Clone: MU
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR