Tisk…

Anti-DUT Rabbit Polyclonal Antibody

Dodavatel: Novus Biologicals
NB100-56624
NOVUNB100-56624EA 12360 CZK
NOVUNB100-56624
Anti-DUT Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: DUT (deoxyuridine triphosphatase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR