Tisk…

Anti-EPAS1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: ep190b]

Dodavatel: Novus Biologicals
NB100-132PE
NOVUNB100-132PEEA 22950 CZK
NOVUNB100-132PE
Anti-EPAS1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: ep190b]
Protilátky
Type: Primary
Antigen: EPAS1 (endothelial PAS domain protein 1)
Clonality: monoclonal
Clone: ep190b
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR