Tisk…
Beakers, low form
Dodavatel: NICKEL ELECTRO
201 202 203 204 206 205 207 209 207A
NICK201EA 2270 CZK
NICK201 NICK202 NICK203 NICK204 213-0264 NICK205 213-0265 213-0266 NICK207A
Beakers, low form
Kádinky
Stainless steel, rust-proof, with spout.
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.