Tisk…
Chemical resistant tape, Kappler™ ChemTape®
Dodavatel: NEW PIG
WPL591
NEWPWPL591EA 1470 CZK
NEWPWPL591
Chemical resistant tape, Kappler™ ChemTape®
Pásky
Seam-sealing chem tape designed and tested for use with chemical-resistant clothing.
  • Tears easily by hand
  • Protects better than duct tape
  • Excellent for securing gloves and boots to protective clothing
Ideal for haz-mat response, this tape is designed and tested to ensure chemical resistance.