Tisk…
Adjustable Hanging Straps for Leak Diverters
Dodavatel: NEW PIG
TLS559
NEWPTLS559EA 0 CZK
NEWPTLS559
Adjustable Hanging Straps for Leak Diverters
Ochrana před pádem Postroj celotělový bezpečnostní
Hang the PIG® leak diverters from uneven surfaces using these adjustable hanging straps.
  • Adjustable tension and length offer a variety of uses
  • Co-polymer hook design is up to five times stronger than most rubber cord hooks
  • Won't scratch paint or finish
  • Premium rubber cord is strong and durable
These hanging straps are made of 100% nylon with premium rubber compound core and co-polymer hooks.