Tisk…
Manual syphon pump
Dodavatel: NEW PIG
PMPL105 PMPL100 PMPE104
NEWPPMPL105EA 414 CZK
NEWPPMPL105 NEWPPMPL100 NEWPPMPE104
Manual syphon pump
Čerpadla Pumpy sudové
Save time and reduce waste with this chemical-resistant syphon pump.
  • Durable polyethylene construction resists chemicals to maximise life of pump
  • For water based liquids and general-purpose chemicals
  • 380 mm long suction tube is ideal for smaller containers or drums up to 25 litres
  • 534 mm delivery hose for accurate dispensing
  • Easy-to-use manual suction operation; an economical and light-weight solution for every budget
It delivers mild solvents, light oils, corrosives, dilute acid/caustics, liquid waxes and disinfectants at up to 20 L/min.